Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGATGACTCCTTGCTGGAGCTGTC[A/G]GAGGGAGAAGAAGATGATGGTGATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611889 MIM: 603623 | ||||||||||||||||||||
Literature Links: |
S100PBP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
S100PBP - S100P binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256121.1 | 352 | Silent Mutation | TCA,TCG | S,S 42 | NP_001243050.1 | |
NM_022753.3 | 352 | Silent Mutation | TCA,TCG | S,S 42 | NP_073590.2 | |
XM_011541961.2 | 352 | Silent Mutation | TCA,TCG | S,S 42 | XP_011540263.1 | |
XM_011541962.2 | 352 | Silent Mutation | TCA,TCG | S,S 42 | XP_011540264.1 | |
XM_011541963.1 | 352 | Silent Mutation | TCA,TCG | S,S 42 | XP_011540265.1 | |
XM_017002098.1 | 352 | Silent Mutation | TCA,TCG | S,S 42 | XP_016857587.1 |
YARS - tyrosyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |