Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTGGACTGAGGAGAGGAGGGGGA[G/T]CTGGCCCTTTAGGTACTGTGGTGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605457 MIM: 616832 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL22RA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL22RA1 - interleukin 22 receptor subunit alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021258.3 | 1689 | Missense Mutation | NP_067081.2 | |||
XM_011541882.1 | 1689 | Missense Mutation | XP_011540184.1 |
MYOM3 - myomesin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |