Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCAAAGAAGAAGAAAAAGAAGAA[A/G]AAGCACAAGAAGAAGCACAAGGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 134651 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FABP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FABP3 - fatty acid binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320996.1 | 992 | Intron | NP_001307925.1 | |||
NM_004102.4 | 992 | Intron | NP_004093.1 | |||
XM_011541007.2 | 992 | Intron | XP_011539309.1 |
ZCCHC17 - zinc finger CCHC-type containing 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282566.1 | 992 | Silent Mutation | AAA,AAG | K,K 255 | NP_001269495.1 | |
NM_001282567.1 | 992 | Silent Mutation | AAA,AAG | K,K 235 | NP_001269496.1 | |
NM_001282568.1 | 992 | Silent Mutation | AAA,AAG | K,K 233 | NP_001269497.1 | |
NM_001282569.1 | 992 | Silent Mutation | AAA,AAG | K,K 225 | NP_001269498.1 | |
NM_001282570.1 | 992 | Silent Mutation | AAA,AAG | K,K 209 | NP_001269499.1 | |
NM_001282571.1 | 992 | Silent Mutation | AAA,AAG | K,K 171 | NP_001269500.1 | |
NM_001282572.1 | 992 | Missense Mutation | AAA,AGA | K,R 151 | NP_001269501.1 | |
NM_001282573.1 | 992 | Missense Mutation | AAA,AGA | K,R 143 | NP_001269502.1 | |
NM_001282574.1 | 992 | UTR 3 | NP_001269503.1 | |||
NM_016505.3 | 992 | Silent Mutation | AAA,AAG | K,K 233 | NP_057589.2 | |
XM_006710681.3 | 992 | Silent Mutation | AAA,AAG | K,K 225 | XP_006710744.1 | |
XM_011541567.2 | 992 | Silent Mutation | AAA,AAG | K,K 233 | XP_011539869.1 |