Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAACCAGCGCTCGGGCATGAAGG[C/T]AGAGAAGAAGGCAGCAGCCAGCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607779 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AUNIP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AUNIP - aurora kinase A and ninein interacting protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PAQR7 - progestin and adipoQ receptor family member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178422.5 | 1258 | Missense Mutation | ACC,GCC | T,A 257 | NP_848509.1 | |
XM_005245745.3 | 1258 | Missense Mutation | ACC,GCC | T,A 257 | XP_005245802.1 | |
XM_005245746.3 | 1258 | Missense Mutation | ACC,GCC | T,A 257 | XP_005245803.1 | |
XM_011540861.1 | 1258 | Missense Mutation | ACC,GCC | T,A 257 | XP_011539163.1 | |
XM_011540862.2 | 1258 | Missense Mutation | ACC,GCC | T,A 257 | XP_011539164.1 |