Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600114 MIM: 610962 MIM: 615531 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCT3 - chaperonin containing TCP1 subunit 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GLMP - glycosylated lysosomal membrane protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMG5 - SMG5, nonsense mediated mRNA decay factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM79 - transmembrane protein 79 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VHLL - von Hippel-Lindau tumor suppressor like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004319.2 | 597 | Missense Mutation | TCA,TTA | S,L 50 | NP_001004319.1 |