Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCTCCTCACAGGATCTTCCAAA[C/T]GTTGGAAAGGAACCAACCCTAATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 187040 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TAL1 - TAL bHLH transcription factor 1, erythroid differentiation factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287347.2 | 3172 | UTR 3 | NP_001274276.1 | |||
NM_001290403.1 | 3172 | UTR 3 | NP_001277332.1 | |||
NM_001290404.1 | 3172 | UTR 3 | NP_001277333.1 | |||
NM_001290405.1 | 3172 | UTR 3 | NP_001277334.1 | |||
NM_001290406.1 | 3172 | UTR 3 | NP_001277335.1 | |||
NM_003189.5 | 3172 | UTR 3 | NP_003180.1 | |||
XM_005271160.4 | 3172 | UTR 3 | XP_005271217.1 | |||
XM_017002187.1 | 3172 | UTR 3 | XP_016857676.1 | |||
XM_017002188.1 | 3172 | UTR 3 | XP_016857677.1 | |||
XM_017002189.1 | 3172 | UTR 3 | XP_016857678.1 | |||
XM_017002190.1 | 3172 | UTR 3 | XP_016857679.1 | |||
XM_017002191.1 | 3172 | UTR 3 | XP_016857680.1 | |||
XM_017002192.1 | 3172 | Intron | XP_016857681.1 | |||
XM_017002193.1 | 3172 | UTR 3 | XP_016857682.1 |