Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGCACCCCAATCCTCTTGATGTC[A/G]CTGTGGGCCGGGAGGGAGAGAGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 176946 | ||||||||||||||||||||
Literature Links: |
EPHA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EPHA2 - EPH receptor A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017000534.1 | 2904 | Silent Mutation | GAC,GAT | D,D 939 | XP_016856023.1 | |
XM_017000535.1 | 2904 | Silent Mutation | GAC,GAT | D,D 931 | XP_016856024.1 | |
XM_017000536.1 | 2904 | Silent Mutation | GAC,GAT | D,D 888 | XP_016856025.1 | |
XM_017000537.1 | 2904 | Intron | XP_016856026.1 |