Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCACCCAGGCCAAGAGAACAGGG[C/G]CATAGACAGCCGTCTGGACTGGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 614264 MIM: 616041 MIM: 191523 | ||||||||||||||||||||
Literature Links: |
ARHGAP30 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGAP30 - Rho GTPase activating protein 30 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025598.1 | 2548 | Silent Mutation | TGC,TGT | C,C 911 | NP_001020769.1 | |
NM_001287600.1 | 2548 | Silent Mutation | TGC,TGT | C,C 763 | NP_001274529.1 | |
NM_001287602.1 | 2548 | Silent Mutation | TGC,TGT | C,C 734 | NP_001274531.1 | |
NM_181720.2 | 2548 | Silent Mutation | TGC,TGT | C,C 700 | NP_859071.2 | |
XM_005245070.2 | 2548 | Silent Mutation | TGC,TGT | C,C 854 | XP_005245127.1 | |
XM_005245073.3 | 2548 | Silent Mutation | TGC,TGT | C,C 763 | XP_005245130.1 | |
XM_011509391.2 | 2548 | Silent Mutation | TGC,TGT | C,C 763 | XP_011507693.1 | |
XM_017000960.1 | 2548 | Silent Mutation | TGC,TGT | C,C 777 | XP_016856449.1 |
TSTD1 - thiosulfate sulfurtransferase like domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USF1 - upstream transcription factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |