Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCCTAGCACACCCAGGGCTGCCC[G/T]CACAGCACGGCTGAGCAGTGAGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604468 MIM: 608042 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP3K6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP3K6 - mitogen-activated protein kinase kinase kinase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297609.1 | 3582 | Silent Mutation | NP_001284538.1 | |||
NM_004672.4 | 3582 | Silent Mutation | NP_004663.3 | |||
XM_017002771.1 | 3582 | Missense Mutation | XP_016858260.1 | |||
XM_017002772.1 | 3582 | Intron | XP_016858261.1 |
SYTL1 - synaptotagmin like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |