Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGGTCTATTTCATGAGAATATG[C/T]GTAGACCATACTTTCCAGTAGCTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605336 | ||||||||||||||||||||
Literature Links: |
CFHR3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CFHR3 - complement factor H related 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166624.1 | 202 | Missense Mutation | CGT,TGT | R,C 39 | NP_001160096.1 | |
NM_021023.5 | 202 | Missense Mutation | CGT,TGT | R,C 39 | NP_066303.2 |