Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGCACTCTGCATGTGACTCCCC[A/G]CAGCCTGGACAGGTGAGTGACTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LAX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LAX1 - lymphocyte transmembrane adaptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136190.1 | 378 | Intron | NP_001129662.1 | |||
NM_001282878.1 | 378 | Intron | NP_001269807.1 | |||
NM_017773.3 | 378 | Missense Mutation | CAC,CGC | H,R 26 | NP_060243.2 | |
XM_006711397.3 | 378 | Missense Mutation | CAC,CGC | H,R 26 | XP_006711460.1 |