Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAGCAGGCACACCCAGAGGGCCT[C/T]GGCACCTCATTGCTGGTGGCACCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 613535 MIM: 608458 | ||||||||||||||||||||
Literature Links: |
KIAA0319L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIAA0319L - KIAA0319 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NCDN - neurochondrin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014839.1 | 567 | Missense Mutation | CGG,TGG | R,W 170 | NP_001014839.1 | |
NM_001014841.1 | 567 | Missense Mutation | CGG,TGG | R,W 153 | NP_001014841.1 | |
NM_014284.2 | 567 | Missense Mutation | CGG,TGG | R,W 170 | NP_055099.1 |