Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCATCTGGTCCTCCGTCTCCACCC[A/G]GCTCCCTGCCCCAGGACCTTCCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 601738 | ||||||||||||||||||||
Literature Links: |
EXTL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EXTL1 - exostosin like glycosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004455.2 | 1396 | Silent Mutation | CCA,CCG | P,P 166 | NP_004446.2 | |
XM_005245779.3 | 1396 | Silent Mutation | CCA,CCG | P,P 166 | XP_005245836.1 | |
XM_017000650.1 | 1396 | Silent Mutation | CCA,CCG | P,P 166 | XP_016856139.1 |