Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGAAAAGTTCAACCATTTTTCC[A/G]AAGATAAACAGCTAGTGAGTGGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600234 MIM: 606879 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMGCS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HMGCS2 - 3-hydroxy-3-methylglutaryl-CoA synthase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166107.1 | Intron | NP_001159579.1 | ||||
NM_005518.3 | Intron | NP_005509.1 | ||||
XM_011541313.1 | Intron | XP_011539615.1 |
PHGDH - phosphoglycerate dehydrogenase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |