Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGCAGCCACAGCCAGCATAAAGA[C/T]AATGAAGACATTCTTCTCTGTGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 121013 | ||||||||||||||||||||
Literature Links: |
GJA5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GJA5 - gap junction protein alpha 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005266.6 | 2808 | Missense Mutation | ATC,GTC | I,V 209 | NP_005257.2 | |
NM_181703.3 | 2808 | Missense Mutation | ATC,GTC | I,V 209 | NP_859054.1 | |
XM_005272951.4 | 2808 | Missense Mutation | ATC,GTC | I,V 209 | XP_005273008.1 | |
XM_017001044.1 | 2808 | Missense Mutation | ATC,GTC | I,V 209 | XP_016856533.1 |