Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGGGCCCATCTACATGCGAGAA[A/G]ATGGGCAGCTGCACATGGTCAATCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
HSPB11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HSPB11 - heat shock protein family B (small) member 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC42 - leucine rich repeat containing 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256409.1 | 155 | Missense Mutation | AAT,GAT | N,D 21 | NP_001243338.1 | |
NM_052940.4 | 155 | Missense Mutation | AAT,GAT | N,D 21 | NP_443172.1 | |
XM_006710327.3 | 155 | Missense Mutation | AAT,GAT | N,D 21 | XP_006710390.1 | |
XM_006710328.3 | 155 | Missense Mutation | AAT,GAT | N,D 21 | XP_006710391.1 |