Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAATGGAAAAGCAAAAGCCCTT[C/T]GCATTGTTCGTACCACCGAGATCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602162 | ||||||||||||||||||||
Literature Links: |
SYCP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SYCP1 - synaptonemal complex protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282541.1 | 264 | Silent Mutation | TTC,TTT | F,F 7 | NP_001269470.1 | |
NM_001282542.1 | 264 | Silent Mutation | TTC,TTT | F,F 7 | NP_001269471.1 | |
NM_003176.3 | 264 | Silent Mutation | TTC,TTT | F,F 7 | NP_003167.2 | |
XM_005271155.3 | 264 | Silent Mutation | TTC,TTT | F,F 7 | XP_005271212.1 | |
XM_005271156.1 | 264 | Silent Mutation | TTC,TTT | F,F 7 | XP_005271213.1 | |
XM_006710859.1 | 264 | Silent Mutation | TTC,TTT | F,F 7 | XP_006710922.1 | |
XM_011542037.2 | 264 | Silent Mutation | TTC,TTT | F,F 7 | XP_011540339.1 | |
XM_011542038.2 | 264 | Intron | XP_011540340.1 | |||
XM_011542039.1 | 264 | Silent Mutation | TTC,TTT | F,F 7 | XP_011540341.1 | |
XM_017002184.1 | 264 | Silent Mutation | TTC,TTT | F,F 7 | XP_016857673.1 | |
XM_017002185.1 | 264 | Intron | XP_016857674.1 |