Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCAGAGGGTTGAAGGCATAGAA[A/C]CTCAGTCCAAAGTGCCTGAGGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608477 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKR7A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKR7A3 - aldo-keto reductase family 7 member A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012067.2 | 390 | Missense Mutation | NP_036199.2 | |||
XM_011541046.2 | 390 | Missense Mutation | XP_011539348.1 | |||
XM_017000714.1 | 390 | Missense Mutation | XP_016856203.1 |
LOC100506730 - uncharacterized LOC100506730 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |