Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTAAATTGATCTTTAATGTTAGT[A/G]TTAATATAGAATGGTCCATACCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYDE2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SYDE2 - synapse defective Rho GTPase homolog 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032184.1 | Intron | NP_115560.1 | ||||
XM_005271254.3 | Intron | XP_005271311.1 | ||||
XM_006710946.3 | Intron | XP_006711009.1 | ||||
XM_017002483.1 | Intron | XP_016857972.1 | ||||
XM_017002484.1 | Intron | XP_016857973.1 | ||||
XM_017002485.1 | Intron | XP_016857974.1 | ||||
XM_017002486.1 | Intron | XP_016857975.1 |