Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGACTCACAGCTGAAGGGGACAGC[C/T]CTCCAGCTCCTCCAGGACCCCGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 147671 MIM: 191315 | ||||||||||||||||||||
Literature Links: |
INSRR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
INSRR - insulin receptor related receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014215.2 | 3945 | Missense Mutation | AGC,GGC | S,G 1216 | NP_055030.1 |
NTRK1 - neurotrophic receptor tyrosine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007792.1 | 3945 | Intron | NP_001007793.1 | |||
NM_001012331.1 | 3945 | Intron | NP_001012331.1 | |||
NM_002529.3 | 3945 | Intron | NP_002520.2 |