Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTACTCAGGGCTGCTGGGGGTGGG[C/T]GATGAGATCCTCGAGGTGAACGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603944 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA1614 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA1614 - KIAA1614 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STX6 - syntaxin 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286210.1 | 1260 | Intron | NP_001273139.1 | |||
NM_005819.5 | 1260 | Intron | NP_005810.1 | |||
XM_011509058.1 | 1260 | UTR 3 | XP_011507360.1 | |||
XM_011509059.1 | 1260 | UTR 3 | XP_011507361.1 | |||
XM_017000006.1 | 1260 | UTR 3 | XP_016855495.1 | |||
XM_017000007.1 | 1260 | Intron | XP_016855496.1 |