Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGACAGCCACGGTCAGGGCCATG[C/T]TAAGGGGAATGTCAGCATCGGACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605037 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIF17 - kinesin family member 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH2D5 - SH2 domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001103160.1 | 2301 | UTR 3 | NP_001096630.1 | |||
NM_001103161.1 | 2301 | UTR 3 | NP_001096631.1 | |||
XM_011541459.1 | 2301 | UTR 3 | XP_011539761.1 | |||
XM_011541460.1 | 2301 | UTR 3 | XP_011539762.1 | |||
XM_011541461.2 | 2301 | UTR 3 | XP_011539763.1 | |||
XM_011541462.1 | 2301 | UTR 3 | XP_011539764.1 |