Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCTTCAAGTTCAGAAGATGCAGG[A/G]TTTTTTTCCTGTTCTTCCATTTTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612208 MIM: 609824 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GOLPH3L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GOLPH3L - golgi phosphoprotein 3 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HORMAD1 - HORMA domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199829.1 | 946 | Silent Mutation | NP_001186758.1 | |||
NM_032132.4 | 946 | Silent Mutation | NP_115508.2 | |||
XM_006711570.2 | 946 | Silent Mutation | XP_006711633.1 | |||
XM_006711571.2 | 946 | Silent Mutation | XP_006711634.1 | |||
XM_011510053.2 | 946 | Missense Mutation | XP_011508355.1 | |||
XM_011510054.2 | 946 | Silent Mutation | XP_011508356.1 | |||
XM_011510055.2 | 946 | Silent Mutation | XP_011508357.1 | |||
XM_011510056.2 | 946 | Silent Mutation | XP_011508358.1 |