Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATATTATGGAATTCTGACCTCTA[G/T]AAGACAAGTGATAATACAATATGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615734 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLCC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLCC1 - chloride channel CLIC like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR47 - WD repeat domain 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142550.1 | 3621 | UTR 3 | NP_001136022.1 | |||
NM_001142551.1 | 3621 | UTR 3 | NP_001136023.1 | |||
NM_014969.5 | 3621 | UTR 3 | NP_055784.3 | |||
XM_006710460.3 | 3621 | UTR 3 | XP_006710523.1 | |||
XM_011541027.2 | 3621 | UTR 3 | XP_011539329.1 | |||
XM_011541028.2 | 3621 | UTR 3 | XP_011539330.1 | |||
XM_011541029.1 | 3621 | UTR 3 | XP_011539331.1 | |||
XM_011541030.1 | 3621 | UTR 3 | XP_011539332.1 | |||
XM_011541031.1 | 3621 | UTR 3 | XP_011539333.1 | |||
XM_017000696.1 | 3621 | UTR 3 | XP_016856185.1 | |||
XM_017000697.1 | 3621 | UTR 3 | XP_016856186.1 |