Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTTCTGCACCTTCATGATTTCAT[C/T]CTCCTTTTGGGAAGTGAGGATGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603594 | ||||||||||||||||||||
Literature Links: |
TNFSF4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TNFSF4 - tumor necrosis factor superfamily member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297562.1 | 871 | Missense Mutation | AAT,GAT | N,D 33 | NP_001284491.1 | |
NM_003326.4 | 871 | Missense Mutation | AAT,GAT | N,D 83 | NP_003317.1 | |
XM_011509964.2 | 871 | Missense Mutation | AAT,GAT | N,D 179 | XP_011508266.2 | |
XM_017002228.1 | 871 | Missense Mutation | AAT,GAT | N,D 115 | XP_016857717.1 | |
XM_017002229.1 | 871 | Missense Mutation | AAT,GAT | N,D 94 | XP_016857718.1 | |
XM_017002230.1 | 871 | Missense Mutation | AAT,GAT | N,D 92 | XP_016857719.1 |