Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCATTCTTTTCATGTTCGTTACA[A/G]GTAAGACTATTGCTGTCTTAATGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609245 | ||||||||||||||||||||
Literature Links: |
GPSM2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPSM2 - G-protein signaling modulator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321038.1 | 552 | Missense Mutation | AAA,AGA | K,R 19 | NP_001307967.1 | |
NM_001321039.1 | 552 | Missense Mutation | AAA,AGA | K,R 19 | NP_001307968.1 | |
NM_013296.4 | 552 | Missense Mutation | AAA,AGA | K,R 19 | NP_037428.3 | |
XM_006710589.2 | 552 | Intron | XP_006710652.1 | |||
XM_011541301.2 | 552 | Missense Mutation | AAA,AGA | K,R 19 | XP_011539603.1 | |
XM_011541302.2 | 552 | Missense Mutation | AAA,AGA | K,R 19 | XP_011539604.1 | |
XM_011541303.2 | 552 | Missense Mutation | AAA,AGA | K,R 19 | XP_011539605.1 | |
XM_017001097.1 | 552 | Missense Mutation | AAA,AGA | K,R 19 | XP_016856586.1 | |
XM_017001098.1 | 552 | Missense Mutation | AAA,AGA | K,R 19 | XP_016856587.1 |