Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGCAGGAGCTCAAGGACCTGGGC[A/G]TCACCGTGTTCATTGTCAGCACCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611901 | ||||||||||||||||||||
Literature Links: |
LOC102724312 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC102724312 - uncharacterized LOC102724312 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC107985729 - uncharacterized LOC107985729 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM88B - transmembrane protein 88B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VWA1 - von Willebrand factor A domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022834.4 | 318 | Missense Mutation | ATC,GTC | I,V 163 | NP_073745.2 | |
NM_199121.2 | 318 | Missense Mutation | CAT,CGT | H,R 31 | NP_954572.2 |