Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACGTTCGGGCTCATCTTTTACTT[C/T]TCCACGGACAACCTGGTGCGGCCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600759 | ||||||||||||||||||||
Literature Links: |
LOC105373120 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105373120 - uncharacterized LOC105373120 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSEN2 - presenilin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000447.2 | 1521 | Silent Mutation | TTC,TTT | F,F 428 | NP_000438.2 | |
NM_012486.2 | 1521 | Silent Mutation | TTC,TTT | F,F 427 | NP_036618.2 | |
XM_005273199.3 | 1521 | Silent Mutation | TTC,TTT | F,F 428 | XP_005273256.1 | |
XM_017001835.1 | 1521 | Silent Mutation | TTC,TTT | F,F 428 | XP_016857324.1 | |
XM_017001836.1 | 1521 | Silent Mutation | TTC,TTT | F,F 427 | XP_016857325.1 |