Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGACTCAGGCCCTCTTCTCTGCTT[A/G]GCTGGATTATTTCATTACTCCACTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153420 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD58 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD58 - CD58 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144822.1 | Intron | NP_001138294.1 | ||||
NM_001779.2 | Intron | NP_001770.1 | ||||
XM_017002869.1 | Intron | XP_016858358.1 |