Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCACCTCCCTGGGGGTCACGGCCA[A/T]CCCCTTTTGTGCCTGCAGGTAGTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609859 MIM: 604748 | ||||||||||||||||||||
Literature Links: |
ETNK2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ETNK2 - ethanolamine kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297760.1 | 1285 | Missense Mutation | ATG,TTG | M,L 319 | NP_001284689.1 | |
NM_001297761.1 | 1285 | Missense Mutation | ATG,TTG | M,L 141 | NP_001284690.1 | |
NM_001297762.1 | 1285 | Missense Mutation | ATG,TTG | M,L 278 | NP_001284691.1 | |
NM_018208.3 | 1285 | Missense Mutation | ATG,TTG | M,L 319 | NP_060678.2 | |
XM_011509714.2 | 1285 | Missense Mutation | ATG,TTG | M,L 141 | XP_011508016.1 | |
XM_011509715.1 | 1285 | Missense Mutation | ATG,TTG | M,L 141 | XP_011508017.1 | |
XM_017001632.1 | 1285 | Missense Mutation | ATG,TTG | M,L 141 | XP_016857121.1 |
LOC101929441 - uncharacterized LOC101929441 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SOX13 - SRY-box 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |