Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTCCAGAACCACACCTGCCTGAG[A/G]AAGGGGAAGGGGGTAAGCCTTGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609017 | ||||||||||||||||||||
Literature Links: |
ERMAP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ERMAP - erythroblast membrane associated protein (Scianna blood group) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC339539 - uncharacterized LOC339539 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF691 - zinc finger protein 691 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242739.1 | 4123 | Missense Mutation | AAA,GAA | K,E 47 | NP_001229668.1 | |
NM_015911.3 | 4123 | Missense Mutation | AAA,GAA | K,E 16 | NP_056995.1 | |
XM_006710671.3 | 4123 | Missense Mutation | AAA,GAA | K,E 43 | XP_006710734.1 | |
XM_017001400.1 | 4123 | Missense Mutation | AAA,GAA | K,E 47 | XP_016856889.1 | |
XM_017001401.1 | 4123 | Missense Mutation | AAA,GAA | K,E 47 | XP_016856890.1 | |
XM_017001402.1 | 4123 | Missense Mutation | AAA,GAA | K,E 38 | XP_016856891.1 | |
XM_017001403.1 | 4123 | Missense Mutation | AAA,GAA | K,E 38 | XP_016856892.1 |