Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATCTCACACTCATTCTCCAGCCG[C/T]TCCAGTTTCAGGTGTGCTGTGCCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603755 | ||||||||||||||||||||
Literature Links: |
CC2D1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CC2D1B - coiled-coil and C2 domain containing 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032449.2 | 2628 | Silent Mutation | GAA,GAG | E,E 803 | NP_115825.1 | |
XM_005270590.2 | 2628 | Silent Mutation | GAA,GAG | E,E 803 | XP_005270647.1 | |
XM_005270591.2 | 2628 | Silent Mutation | GAA,GAG | E,E 797 | XP_005270648.1 | |
XM_005270593.2 | 2628 | Silent Mutation | GAA,GAG | E,E 494 | XP_005270650.1 | |
XM_006710423.2 | 2628 | Silent Mutation | GAA,GAG | E,E 761 | XP_006710486.1 | |
XM_017000559.1 | 2628 | Silent Mutation | GAA,GAG | E,E 488 | XP_016856048.1 |
ZFYVE9 - zinc finger FYVE-type containing 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |