Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTAGTTCAAAAATAATCTGTAA[G/T]TGCTGTAAGAAATGTCAACCACTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600413 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GBP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GBP3 - guanylate binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319179.1 | 2924 | UTR 3 | NP_001306108.1 | |||
NM_001319180.1 | 2924 | UTR 3 | NP_001306109.1 | |||
NM_001319181.1 | 2924 | Intron | NP_001306110.1 | |||
NM_018284.2 | 2924 | UTR 3 | NP_060754.2 | |||
XM_005270743.2 | 2924 | UTR 3 | XP_005270800.1 | |||
XM_006710574.2 | 2924 | UTR 3 | XP_006710637.1 | |||
XM_011541233.1 | 2924 | UTR 3 | XP_011539535.1 | |||
XM_011541234.1 | 2924 | UTR 3 | XP_011539536.1 | |||
XM_017001013.1 | 2924 | UTR 3 | XP_016856502.1 | |||
XM_017001014.1 | 2924 | UTR 3 | XP_016856503.1 |