Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGAAAGCAAGGGTTTGGCCAGT[C/T]AGGGATGAGGATGGGGCCCGAGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176946 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPHA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPHA2 - EPH receptor A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017000534.1 | 3519 | UTR 3 | XP_016856023.1 | |||
XM_017000535.1 | 3519 | UTR 3 | XP_016856024.1 | |||
XM_017000536.1 | 3519 | UTR 3 | XP_016856025.1 | |||
XM_017000537.1 | 3519 | Intron | XP_016856026.1 |