Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCTTTTCAGGGAAGGAGATCAC[A/G]TTGCTGATGCAGACATTGAATACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610563 MIM: 608676 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KPNA6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KPNA6 - karyopherin subunit alpha 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TXLNA - taxilin alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175852.3 | 438 | Silent Mutation | ACA,ACG | T,T 173 | NP_787048.1 | |
XM_011540931.2 | 438 | Intron | XP_011539233.1 | |||
XM_011540932.2 | 438 | Intron | XP_011539234.1 | |||
XM_017000561.1 | 438 | Silent Mutation | ACA,ACG | T,T 206 | XP_016856050.1 | |
XM_017000562.1 | 438 | Silent Mutation | ACA,ACG | T,T 173 | XP_016856051.1 | |
XM_017000563.1 | 438 | Silent Mutation | ACA,ACG | T,T 206 | XP_016856052.1 | |
XM_017000564.1 | 438 | UTR 5 | XP_016856053.1 | |||
XM_017000565.1 | 438 | UTR 5 | XP_016856054.1 |