Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCCCATGTGCTCCACAGAGGCCA[A/G]CCCGTGGCGCTGCCTATCATTGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPOCD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPOCD1 - SPOC domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281987.1 | 2785 | Silent Mutation | CTG,TTG | L,L 958 | NP_001268916.1 | |
NM_001281988.1 | 2785 | Silent Mutation | CTG,TTG | L,L 451 | NP_001268917.1 | |
NM_144569.5 | 2785 | Silent Mutation | CTG,TTG | L,L 958 | NP_653170.3 | |
XM_011542413.2 | 2785 | Silent Mutation | CTG,TTG | L,L 451 | XP_011540715.1 | |
XM_011542414.2 | 2785 | Silent Mutation | CTG,TTG | L,L 451 | XP_011540716.1 | |
XM_017002779.1 | 2785 | Silent Mutation | CTG,TTG | L,L 957 | XP_016858268.1 | |
XM_017002780.1 | 2785 | Silent Mutation | CTG,TTG | L,L 944 | XP_016858269.1 | |
XM_017002781.1 | 2785 | Silent Mutation | CTG,TTG | L,L 916 | XP_016858270.1 | |
XM_017002782.1 | 2785 | Silent Mutation | CTG,TTG | L,L 701 | XP_016858271.1 |