Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTTATCACAAGACACCTTCCCT[C/G]CATGAGGTTAGTACCTTGGTTTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607356 | ||||||||||||||||||||
Literature Links: |
AGO4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AGO4 - argonaute 4, RISC catalytic component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017629.3 | 557 | Missense Mutation | TCC,TGC | S,C 161 | NP_060099.2 | |
XM_005270578.3 | 557 | Missense Mutation | TCC,TGC | S,C 94 | XP_005270635.1 | |
XM_005270579.3 | 557 | Missense Mutation | TCC,TGC | S,C 94 | XP_005270636.1 | |
XM_011540883.2 | 557 | UTR 5 | XP_011539185.1 | |||
XM_011540884.2 | 557 | UTR 5 | XP_011539186.1 | |||
XM_017000529.1 | 557 | Missense Mutation | TCC,TGC | S,C 161 | XP_016856018.1 |