Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGAGGCAGTCATAGTCCTCAGA[C/G]GCCCCAGCTCCATTCCCCAGCTCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616262 MIM: 606225 MIM: 165270 | ||||||||||||||||||||
Literature Links: |
KLHL21 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLHL21 - kelch like family member 21 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAS1R1 - taste 1 receptor member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB48 - zinc finger and BTB domain containing 48 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278647.1 | 670 | Missense Mutation | ACG,AGG | T,R 174 | NP_001265576.1 | |
NM_001278648.1 | 670 | Missense Mutation | ACG,AGG | T,R 174 | NP_001265577.1 | |
NM_005341.3 | 670 | Missense Mutation | ACG,AGG | T,R 174 | NP_005332.1 | |
XM_017001110.1 | 670 | Missense Mutation | ACG,AGG | T,R 174 | XP_016856599.1 | |
XM_017001111.1 | 670 | Missense Mutation | ACG,AGG | T,R 174 | XP_016856600.1 | |
XM_017001112.1 | 670 | UTR 5 | XP_016856601.1 | |||
XM_017001113.1 | 670 | UTR 5 | XP_016856602.1 | |||
XM_017001114.1 | 670 | UTR 5 | XP_016856603.1 |