Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTTGTGATGTTACAAAATATAA[C/T]TCCCCAACTAATTTCCAGATAGATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610468 MIM: 613975 | ||||||||||||||||||||
Literature Links: |
IFI44 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IFI44 - interferon induced protein 44 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006417.4 | 1689 | Silent Mutation | AAC,AAT | N,N 109 | NP_006408.3 | |
XM_005270380.3 | 1689 | Missense Mutation | AAC,AAT | N,N 109 | XP_005270437.1 | |
XM_011540516.2 | 1689 | Missense Mutation | AAC,AAT | N,N 109 | XP_011538818.1 | |
XM_017000083.1 | 1689 | Missense Mutation | AAC,AAT | N,N 109 | XP_016855572.1 |
IFI44L - interferon induced protein 44 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |