Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGCGTGTTTAGTTTCGGCTGAAC[A/T]GAATCATTCCTAAAGAAAGTGACTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607718 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYT6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SYT6 - synaptotagmin 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270805.1 | 2340 | UTR 3 | NP_001257734.1 | |||
NM_205848.3 | 2340 | UTR 3 | NP_995320.1 | |||
XM_017000371.1 | 2340 | UTR 3 | XP_016855860.1 |