Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCCTGAGGAAGATGAAGACCAGA[A/G]AAACCCTGCAAAGGAGTCCCTAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 102642 | ||||||||||||||||||||
Literature Links: |
SOAT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SOAT1 - sterol O-acyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252511.1 | 311 | UTR 5 | NP_001239440.1 | |||
NM_001252512.1 | 311 | Intron | NP_001239441.1 | |||
NM_003101.5 | 311 | Missense Mutation | AAA,AGA | K,R 27 | NP_003092.4 | |
XM_011509911.1 | 311 | Missense Mutation | AAA,AGA | K,R 27 | XP_011508213.1 | |
XM_011509912.1 | 311 | Missense Mutation | AAA,AGA | K,R 27 | XP_011508214.1 |