Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTGGGAAGCAGTGTCCATTGAGT[C/T]TTCCTTTCTGGCTTGAAGAGACTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611656 | ||||||||||||||||||||
Literature Links: |
SIKE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SIKE1 - suppressor of IKBKE 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102396.1 | 669 | Missense Mutation | AAC,GAC | N,D 201 | NP_001095866.1 | |
NM_025073.2 | 669 | Missense Mutation | AAC,GAC | N,D 197 | NP_079349.2 |