Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTATATCTTGCTCCTGTTTTTCC[A/G]CAGGATGCACCCTGGACTCCCATCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600246 | ||||||||||||||||||||
Literature Links: |
ELK4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ELK4 - ELK4, ETS transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001973.3 | 1549 | Intron | NP_001964.2 | |||
NM_021795.2 | 1549 | Intron | NP_068567.1 | |||
XM_005244950.4 | 1549 | Intron | XP_005245007.1 | |||
XM_005244951.4 | 1549 | Intron | XP_005245008.1 | |||
XM_017000580.1 | 1549 | Intron | XP_016856069.1 |
MFSD4A - major facilitator superfamily domain containing 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181644.4 | 1549 | Missense Mutation | CAC,CGC | H,R 488 | NP_857595.3 | |
XM_011509229.2 | 1549 | Missense Mutation | CAC,CGC | H,R 401 | XP_011507531.1 |