Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCTTTAATTTTTTATTACAGAAT[C/T]GAAGATGTTCCTGCTCCTTCTACAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603185 | ||||||||||||||||||||
Literature Links: |
NASP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NASP - nuclear autoantigenic sperm protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195193.1 | Intron | NP_001182122.1 | ||||
NM_002482.3 | Intron | NP_002473.2 | ||||
NM_152298.3 | Intron | NP_689511.2 | ||||
XM_005270888.2 | Intron | XP_005270945.1 | ||||
XM_005270889.2 | Intron | XP_005270946.1 | ||||
XM_011541509.1 | Intron | XP_011539811.1 | ||||
XM_017001356.1 | Intron | XP_016856845.1 |