Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCAACTGAGACCATTTAGTGTAA[G/T]GGCGAACAGCCCTCCACCTTTCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603594 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNFSF4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TNFSF4 - tumor necrosis factor superfamily member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297562.1 | 2639 | UTR 3 | NP_001284491.1 | |||
NM_003326.4 | 2639 | UTR 3 | NP_003317.1 | |||
XM_011509964.2 | 2639 | UTR 3 | XP_011508266.2 | |||
XM_017002228.1 | 2639 | UTR 3 | XP_016857717.1 | |||
XM_017002229.1 | 2639 | UTR 3 | XP_016857718.1 | |||
XM_017002230.1 | 2639 | UTR 3 | XP_016857719.1 |