Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTTGTAACAGCCGCTGATAGCTC[A/G]GGCAGTGCTGCTACATCAACCTTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608003 | ||||||||||||||||||||
Literature Links: |
HIPK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HIPK1 - homeodomain interacting protein kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152696.3 | 614 | Silent Mutation | TCA,TCG | S,S 92 | NP_689909.2 | |
NM_181358.2 | 614 | Intron | NP_852003.1 | |||
NM_198268.2 | 614 | Silent Mutation | TCA,TCG | S,S 92 | NP_938009.1 | |
NM_198269.2 | 614 | Intron | NP_938010.1 | |||
XM_005270609.3 | 614 | Silent Mutation | TCA,TCG | S,S 163 | XP_005270666.1 | |
XM_005270610.4 | 614 | Silent Mutation | TCA,TCG | S,S 163 | XP_005270667.1 | |
XM_005270611.4 | 614 | Silent Mutation | TCA,TCG | S,S 163 | XP_005270668.1 | |
XM_005270612.4 | 614 | Silent Mutation | TCA,TCG | S,S 92 | XP_005270669.1 | |
XM_005270613.4 | 614 | Silent Mutation | TCA,TCG | S,S 92 | XP_005270670.1 | |
XM_006710443.3 | 614 | Silent Mutation | TCA,TCG | S,S 163 | XP_006710506.1 | |
XM_011540977.2 | 614 | Silent Mutation | TCA,TCG | S,S 163 | XP_011539279.1 | |
XM_017000605.1 | 614 | Silent Mutation | TCA,TCG | S,S 92 | XP_016856094.1 | |
XM_017000606.1 | 614 | Silent Mutation | TCA,TCG | S,S 163 | XP_016856095.1 |