Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAAAAGCCTCTTCAAAGTACTGC[C/T]GCAGGTTCTGCTTGTTGCTGTCAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608867 | ||||||||||||||||||||
Literature Links: |
DUSP10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DUSP10 - dual specificity phosphatase 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007207.5 | 1390 | Missense Mutation | NP_009138.1 | |||
XM_017000147.1 | 1390 | Intron | XP_016855636.1 |