Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTTCTCCAACTGCTCCTCCAGGC[A/G]CTGTTCAACTCCATGGTTCTCGATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607622 | ||||||||||||||||||||
Literature Links: |
PMVK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PMVK - phosphomevalonate kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323011.1 | 849 | Missense Mutation | CGC,TGC | R,C 162 | NP_001309940.1 | |
NM_001323012.1 | 849 | Missense Mutation | CGC,TGC | R,C 101 | NP_001309941.1 | |
NM_006556.3 | 849 | Missense Mutation | CGC,TGC | R,C 176 | NP_006547.1 |