Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTATCCTCCCCAGAAGGGCTGCAT[C/T]CCCCTCCCGAGGGGTCAGACAGAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 606416 | ||||||||||||||||||||
Literature Links: |
NLRP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NLRP3 - NLR family pyrin domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079821.2 | 863 | Silent Mutation | ATC,ATT | I,I 39 | NP_001073289.1 | |
NM_001127461.2 | 863 | Silent Mutation | ATC,ATT | I,I 39 | NP_001120933.1 | |
NM_001127462.2 | 863 | Silent Mutation | ATC,ATT | I,I 39 | NP_001120934.1 | |
NM_001243133.1 | 863 | Silent Mutation | ATC,ATT | I,I 37 | NP_001230062.1 | |
NM_004895.4 | 863 | Silent Mutation | ATC,ATT | I,I 39 | NP_004886.3 | |
NM_183395.2 | 863 | Silent Mutation | ATC,ATT | I,I 39 | NP_899632.1 | |
XM_011544048.2 | 863 | Silent Mutation | ATC,ATT | I,I 39 | XP_011542350.1 | |
XM_011544053.2 | 863 | Silent Mutation | ATC,ATT | I,I 39 | XP_011542355.1 | |
XM_011544055.2 | 863 | Silent Mutation | ATC,ATT | I,I 39 | XP_011542357.1 | |
XM_017000181.1 | 863 | Silent Mutation | ATC,ATT | I,I 39 | XP_016855670.1 | |
XM_017000182.1 | 863 | Silent Mutation | ATC,ATT | I,I 39 | XP_016855671.1 | |
XM_017000183.1 | 863 | Silent Mutation | ATC,ATT | I,I 39 | XP_016855672.1 | |
XM_017000184.1 | 863 | Silent Mutation | ATC,ATT | I,I 39 | XP_016855673.1 |