Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGAGGAAGCAGCTAGCTGTTAAC[C/T]TCACCCGTGTCCTGGCACTCTACCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611930 MIM: 611836 | ||||||||||||||||||||
Literature Links: |
ISG20L2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ISG20L2 - interferon stimulated exonuclease gene 20 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303095.1 | 268 | Intron | NP_001290024.1 | |||
XM_005245516.4 | 268 | Intron | XP_005245573.1 | |||
XM_005245518.4 | 268 | Intron | XP_005245575.1 | |||
XM_006711560.3 | 268 | Intron | XP_006711623.1 | |||
XM_006711561.3 | 268 | Intron | XP_006711624.1 |
MRPL24 - mitochondrial ribosomal protein L24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RRNAD1 - ribosomal RNA adenine dimethylase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142560.1 | 268 | Missense Mutation | CTC,TTC | L,F 21 | NP_001136032.1 | |
NM_015997.3 | 268 | Missense Mutation | CTC,TTC | L,F 21 | NP_057081.3 | |
XM_005245234.4 | 268 | Missense Mutation | CTC,TTC | L,F 21 | XP_005245291.1 | |
XM_005245235.4 | 268 | Missense Mutation | CTC,TTC | L,F 21 | XP_005245292.1 | |
XM_005245236.4 | 268 | Missense Mutation | CTC,TTC | L,F 21 | XP_005245293.1 | |
XM_005245237.4 | 268 | Missense Mutation | CTC,TTC | L,F 21 | XP_005245294.1 | |
XM_011509600.1 | 268 | Intron | XP_011507902.1 | |||
XM_017001404.1 | 268 | UTR 5 | XP_016856893.1 | |||
XM_017001405.1 | 268 | UTR 5 | XP_016856894.1 | |||
XM_017001406.1 | 268 | UTR 5 | XP_016856895.1 |